Hexamethylenetetramine ≥99.0% for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.18712.0100
8.18712.1000
8.18712.1000
8.18712.5000
CA8.18712.0100EA
59
CAD
CA8.18712.0100
EM8.18712.1000
CA8.18712.1000
CA8.18712.5000
Hexamethylenetetramine ≥99.0% for synthesis, Sigma-Aldrich®
Methenamine
Formula:
C₆H₁₂N₄ Melting Pt: 280 °C Density: 1.33 g/cm3 at 20 °C Flash Pt: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006895 CAS Number: 100-97-0 EINECS: 202-905-8 UN: 1328 ADR: 4.1,III Merck Index: 13,05994 |
|
Specification Test Results
Assay (HClO₄) | ≥99.0% |
Identity (IR) | Passes Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...