1-Octadecanethiol ≥98.0% (by GC), Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: n-Octadecyl mercaptan, n-Octadecanethiol, Stearyl mercaptane, 1-Mercaptooctadecane, Octadecane-1-thiol

M-Clarity™ quality level = MQ200

8.14444.0250 8.14444.0010
CA8.14444.0250EA 151 CAD
CA8.14444.0250 CA8.14444.0010
1-Octadecanethiol ≥98.0% (by GC), Sigma-Aldrich®
1-Octadecanethiol
Formula: C₁₈H₃₈S
Boiling Pt: 366 °C
Melting Pt: 28…31 °C
Density: 0.847 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004886
CAS Number: 2885-00-9
EINECS: 220-744-1
UN: 3335
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR