Polyethylene glycol 400, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
PEG 400
M-Clarity™ quality level = MQ200
8.07485.1000
8.07485.1000
CA8.07485.1000EA
377.24
CAD
CA8.07485.1000
EM8.07485.1000
Polyethylene glycol 400, Sigma-Aldrich®
Polyethylene glycol 400
Formula:
H(OCH₂CH₂)nOH MW: 400 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 4…8 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 245 °C |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...