N,N-Dimethylformamide dimethyl acetal ≥96%, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.03068.0025
8.03068.0001
CA8.03068.0025EA
82
CAD
CA8.03068.0025
CA8.03068.0001
N,N-Dimethylformamide dimethyl acetal ≥96%, Sigma-Aldrich®
N,N-Dimethylformamide dimethyl acetal
Formula:
C₅H₁₃NO₂ Boiling Pt: 102 °C (1013 hPa) Density: 0.893 g/cm³ (20 °C) Flash Pt: 6 °C |
MDL Number:
MFCD00008482 CAS Number: 4637-24-5 EINECS: 225-063-3 UN: 1993 ADR: 3,II |
Specification Test Results
Assay (HClO4-titration) | 96% min |
Density (d 20°/4°) | 0.893-0.897 |
Identity (IR-spectrum) | Conforms |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...