Calcium hydride, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.02100.0100 8.02100.0500 8.02100.0100 8.02100.0500
CA8.02100.0100EA 187 CAD
CA8.02100.0100 CA8.02100.0500 EM8.02100.0100 EM8.02100.0500
Calcium hydride, Sigma-Aldrich®
Calcium hydride
Formula: H₂Ca
Melting Pt: 816 °C
Density: 1.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010897
CAS Number: 7789-78-8
EINECS: 232-189-2
UN: 1404
ADR: 4.3,I
Merck Index: 13,01674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR