Phthalic anhydride, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.00592.1000 8.00592.9050
CA8.00592.1000EA 63 CAD
CA8.00592.1000 CA8.00592.9050
Phthalic anhydride, Sigma-Aldrich®
Phthalic anhydride

8.00592.1000 is offered as an alternative to PX1070-2. Different Pack Size, Same Specifications

Formula: C₈H₄O₃
MW: 148.12 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 131 °C
Density: 1.527 g/cm3 (20 °C)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00005918
CAS Number: 85-44-9
EINECS: 201-607-5
UN: 2214
ADR: 8,III
Merck Index: 13,07457

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR