Propyl-4-hydroxybenzoate sodium salt, EMPROVE® ESSENTIAL BP, Ph. Eur., E212, NF, SAFC®

Supplier: MilliporeSigma
Danger

Synonyms: Propyl parahydroxybenzoate sodium salt, Sodium Propyl Parahydroxybenzoate, Propylparaben Sodium, Propylparaben sodium salt, 4-Hydroxybenzoic acid propyl ester sodium salt, Sodium 4-(propoxycarbonyl)phenolate, Sodium 4-propoxycarbonylphenoxide

1.07428.5000 1.07428.9029
CA1.07428.5000EA 2804.09 CAD
CA1.07428.5000 CA1.07428.9029
Propyl-4-hydroxybenzoate sodium salt, EMPROVE® ESSENTIAL BP, Ph. Eur., E212, NF, SAFC®
Propyl 4-hydroxybenzoate sodium salt
Formula: C₁₀H₁₁NaO₃
MW: 202.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016477
CAS Number: 35285-69-9
EINECS: 252-488-1

Order Now

Specification Test Results

Appearance of solution PASSES
Arsenic (As) 0.0003% max
Butyl-4-hydoxybenzoate 0.2% max
Cadmium (Cd) 0.001% max
Chloride (Cl) 0.03% max
Copper (Cu) 0.001% max
Ethyl-4-hydroxybenzoate 0.3% max
Heavy metals (as Pb) 0.001% max
Identity PASSES
Largest unspecified impurity 0.1% max
Lead (Pb) 0.0005% max
Mercury (Hg) 0.0001% max
Methyl 4-hydroxybenzoate 0.2% max
Organic volatile impurities (according to NF) Conforms
Other Residual Solvents (Ph.Eur./USP) Excluded by manufactoring process
p-Hydroxy Benzoic acid 4.0% max
pH value, 0.1%, water 10.5 max
Related substance (HPLC) Conforms
Residual Solvents (Ph.Eur./USP/ICH) Conforms
Salicylic acid 0.1% max
Sulfate (SO4) 0.03% max
Sulfated ash 34-36%
Sum of peaks (GC) 4.0% max
Water 5.0% max
Zinc (Zn) 0.001% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR