Sodium hydroxide 99 NaOH basis_acidimetry, pellets, EMSURE® ISO ISO LC-MS, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda

MQ300

1.06498.0500 1.06498.1000 1.06498.5000 1.06498.9025 1.06498.9050
CA1.06498.0500CS 1879.17 CAD
CA1.06498.0500 CA1.06498.1000 CA1.06498.5000 CA1.06498.9025 CA1.06498.9050
Sodium hydroxide 99 NaOH basis_acidimetry, pellets, EMSURE® ISO ISO LC-MS, Supelco®
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323 (lit.)
Density: 2.13 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8 II
REACH: 01-2119457892-27
Merck Index: 14,08627

Order Now

Specification Test Results

Assay (acidimetric, NaOH) 99% min
Carbonate (as Na₂CO₃) 1.0% max
Chloride (Cl) 0.012% max
Phosphate (PO₄) 0.0005% max
Silicate (SiO₂) 0.001% max
Sulfate (SO₄) 0.010% max
Total nitrogen (N) 0.0003% max
Heavy metals (as Pb) 0.0005% max
Al (Aluminium) 0.0005% max
As (Aresnic) 0.0001% max
Ca (Calcium) 0.0005% max
Cu (Copper) 0.0002% max
Fe (Iron) 0.0005% max
K (Pottassium) 0.05% max
Mg (Magnesium) 0.0005% max
Ni (Nickel) 0.00025% max
Pb (Lead) 0.0005% max
Zn (Zinc) 0.001% max
. .


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR