L(-)-Menthol, crystals, EMPROVE® ESSENTIAL BP, DAB, Ph. Eur., USP, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Warning

Synonyms: (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, L-Menthol, Levomenthol, L(-)-Menthyl alcoholol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, (1R)-(-)-Menthol, (-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexane, (1R,2S,5R)-(-)-Menthol

1.05995.1000 1.05995.9025
CA1.05995.1000EA 916.21 CAD
CA1.05995.1000 CA1.05995.9025
L(-)-Menthol, crystals, EMPROVE® ESSENTIAL BP, DAB, Ph. Eur., USP, SAFC®
L(-)-Menthol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 41…44 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00062979
CAS Number: 2216-51-5
EINECS: 218-690-9
Merck Index: 13,05861

Order Now

Specification Test Results

Related substances (GC) 1.0% max
Acidity or alkalinity Passes test
Residual Solvents (Ph.Eur./USP/ICH) Excluded by production process
Specific rotation ([a]25/D, 10%, ethanol (96%)) -51--45°
Organic volatile impurities (according to USP) Passes test
Appearance of solution (10%; ethanol 96%) Passes test
Specific rotation ([a]20/D, 10%, ethanol (96%)) -51--48°
Melting range, beginning 41°C min
Melting range, end 44°C max
Assay (GC) 99.0% min
Residue after evaporation 0.05% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR