KIMTECH PURE® G5 Co-Polymer Gloves, Kimberly-Clark
Supplier: Kimberly-Clark
61002 61002 61001 61001 61003 61004 61003 61004
CA32934-040CS 288.77 CAD
32934-040 CA32934-040 32934-042 CA32934-042 32934-044 CA32934-044 32934-038 CA32934-038
KIMTECH PURE® G5 Co-Polymer Gloves, Kimberly-Clark
Gloves
Smooth-finish, vinyl ambidextrous gloves are made via a powderless manufacturing process with proprietary inner coating.
  • They are latex-free and silicone-free to minimize allergic reactions
  • Beaded cuff provides strength for donning
  • AQL 2.5 for pinholes and static dissipative use
Double-bagged with case liner and packaged in a class 100 cleanroom.
Certifications: Recommended for ISO class 5 or higher cleanroom environments.