Metabolomics Column Kits, MicroSolv
Supplier: MicroSolv Technology Corp
43000-05P-2 43000-10P-1 43000-10P-2 43000-15P-2
97039-400EA 2736.48 CAD
97039-400 97039-402 97039-404 97039-406
Metabolomics Column Kits, MicroSolv
Chromatography Columns
Each kit includes one Cogent Bidentate C18™ column and one Cogent Diamond Hydride™ column.
Using these two columns in an LCMS method and the appropriate mass spectrometer, it is easy to create a reliable 'metabolome', which consists of plotting metabolite retention times versus their accurate masses. Using these metabolomes, one can find the differences between sample sets such as healthy cells and diseased cells.
The C18 column is used for the most hydrophobic compounds while the Diamond Hydride™ column is used for all other compounds.