Cogent Bidentate C8™ TYPE-C Silica™ HPLC Columns, MicroSolv
Supplier: MicroSolv Technology Corp
40008-05P 4000875P2
97035-078EA 1659.63 CAD
97035-078 97035-084 97035-086
Cogent Bidentate C8™ TYPE-C Silica™ HPLC Columns, MicroSolv
Chromatography Columns
Using proprietary bonding technology, columns are bonded directly to the nonpolar TYPE-C™ silica surface with two separate points of attachment, allowing them to operate in three different domains: reverse phase, normal phase and aqueous normal phase.
Ordering information: Guard column kits contain five guard columns with or without a holder.
Order Now
SPECIFICATIONS
Environmentally Preferable