Protn-Latex™, G-Biosciences
Supplier: G-Biosciences
786-20LATEX 786-21LATEX
CA82021-356EA 2293.18 CAD
CA82021-356 CA82021-362
Protn-Latex™, G-Biosciences
Assays Protein Assays
A simple and reliable method for estimation of protein contamination in latex
Detects as low as 25-40 μg protein/gm latex (or 25-50 parts per million). Protein solutions applied to the test strips form compact and symmetrical spots, their diameter being directly proportional to protein concentration. Protn-Latex™ latex protein extraction buffer extracts protein from the latex sample, which is then applied to the test strips and rapidly developed. By measuring the diameter of protein spots with the Protn-Latex™ card, the range of protein concentration in the sample can be determined. Available with an optional applicator device.
Ordering information: Accessories available.