Water USP
Supplier: MICROBIX BIOSYSTEMS INC PC-CA
Synonyms:
Hydrogen oxide
WF-60-200
WF-60-020
CA95000-096EA
2771.41
CAD
CA95000-096
CA95000-100
Water USP
Water
Formula:
H₂O Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...