Acetone, EMPROVE® NF, Ph. Eur., BP, SAFC®

Supplier: MilliporeSigma
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

MQ200

1.00013.2500 1.00013.6010 1.00013.6025 1.00013.6190 1.00013.9180 1.00013.9950 1.00013.2500 1.00013.6010 1.00013.6025 1.00013.6190 1.00013.9950
CA1.00013.2500CS 1334.36 CAD
CA1.00013.2500 CA1.00013.6010 CA1.00013.6025 CA1.00013.6190 CA1.00013.9180 CA1.00013.9950 EM1.00013.2500 EM1.00013.6010 EM1.00013.6025 EM1.00013.6190 EM1.00013.9950
Acetone, EMPROVE® NF, Ph. Eur., BP, SAFC®
Acetone

EMPROVE® products are pharmaceutical raw materials with comprehensive documentation

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

2-Propanol (GC) 0.05% max
Acidity or alkalinity Conforms
Appearance of solution Conforms
Benzene (GC) 2ppm max
Cyclohexane (GC) 0.01% max
Density (d 20/20°C) 0.790-0.793
Density (d25/25°C) 0.789 max
Identity (IR) Passes
Identity B (Ph Eur) Passes test
Identity C (Ph Eur) Passes test
Methanol (GC) 0.05% max
Other Impurity (GC) 0.05% max
Other Residual Solvents (Ph.Eur./USP) Excluded by manufacturing process
Purity (GC) 99.0% min
Readily oxidizable substances Passes test
Reducing substances Conforms
Related substances (GC) Conforms
Residue after evaporation 0.004% max
Water (H2O) 0.3% max
Water-insoluble matter Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR