3-Methoxyphenyl isocyanate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24329-06
B24329-14
CAAAB24329-06EA
78.68
CAD
CAAAB24329-06
CAAAB24329-14
CAAAB24329-22
3-Methoxyphenyl isocyanate 99%
3-Methoxyphenyl isocyanate
Purity: 99%
CAS No.: 18908-07-1
Molecular Formula: C8H7NO2
Formula Weight: 149.15
Flash Point: 96°(204°F)
Refractive Index: 1.5430
Odor: Pungent
Boiling Point: 94-95°/10mm
Beilstein Reference: 775986
UN No.: 2206
MDL No.: MFCD00002019
Formula:
C₈H₇NO₂ Boiling Pt: 94…95 °C/10 mm Density: 1.138 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002019 CAS Number: 18908-07-1 EINECS: 242-658-3 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...