2,6-Dimethoxyphenylboronic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24305-03
B24305-06
B24305-14
CAAAB24305-03EA
54.04
CAD
CAAAB24305-03
CAAAB24305-06
CAAAB24305-14
2,6-Dimethoxyphenylboronic acid 97+%
2,6-Dimethoxyphenylboronic acid
Purity: 97+%
CAS No.: 23112-96-1
Molecular Formula: C8H11BO4
Formula Weight: 181.98
Melting Point: 106-108°
Beilstein Reference: 3032352
Formula:
C₈H₁₁BO₄ Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318987 CAS Number: 23112-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...