ω-Bromo-4-phenylacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-4'-phenylacetophenone, Bromomethyl p-phenylphenyl ketone, α-Bromo-4-phenylacetophenone, Biphenyl-4-yl bromomethyl ketone, 4-Phenylphenacyl bromide
B24298-06
B24298-14
B24298-22
CAAAB24298-06EA
59.41
CAD
CAAAB24298-06
CAAAB24298-14
CAAAB24298-22
ω-Bromo-4-phenylacetophenone 97%
ω-Bromo-4-phenylacetophenone
Purity: 97%
CAS No.: 135-73-9
Molecular Formula: C14H11BrO
Formula Weight: 275.15
Melting Point: 123-127°
Beilstein Reference: 513838
UN No.: 3261
MDL No.: MFCD00000202
Formula:
C₁₄H₁₁BrO MW: 275.14 g/mol Boiling Pt: 354 Melting Pt: 123…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000202 CAS Number: 135-73-9 EINECS: 205-217-6 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...