3,4-Dichlorophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B24292-03 B24292-06 B24292-14
CAAAB24292-03EA 24.02 CAD
CAAAB24292-03 CAAAB24292-06 CAAAB24292-14
3,4-Dichlorophenylboronic acid 97%
3,4-Dichlorophenylboronic acid

Purity: 97% CAS No.: 151169-75-4 Molecular Formula: C6H5BCl2O2 Formula Weight: 190.82 Melting Point: 270-274° Beilstein Reference: 7369790

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 270…274 °C
Storage Temperature: Ambient
MDL Number: MFCD01074646
CAS Number: 151169-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR