Ethyl (methylthio)acetate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24285-14
B24285-22
CAAAB24285-06EA
75.22
CAD
CAAAB24285-06
CAAAB24285-14
CAAAB24285-22
Ethyl (methylthio)acetate 98%
Ethyl (methylthio)acetate
Purity: 98%
CAS No.: 4455-13-4
Molecular Formula: CH3SCH2CO2CH2CH3
Formula Weight: 134.20
Flash Point: 59°(138°F)
Refractive Index: 1.4600
Odor: Characteristic
Boiling Point: 176°
Beilstein Reference: 1744999
UN No.: 3272
MDL No.: MFCD00009182
Formula:
C₅H₁₀O₂S Boiling Pt: 176 °C Density: 1.06 Flash Pt: 59 °C(138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009182 CAS Number: 4455-13-4 EINECS: 224-700-2 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...