Dicloran (2,6-dichloro-4-nitroaniline) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B24283-14
B24283-22
B24283-36
CAAAAB24283-14EA
35.55
CAD
CAAAAB24283-14
CAAAB24283-22
CAAAB24283-36
Dicloran (2,6-dichloro-4-nitroaniline) 95%
Dicloran (2,6-Dichloro-4-nitroaniline)
Purity: 95%
CAS No.: 99-30-9
Molecular Formula: C6H4Cl2N2O2
Formula Weight: 207.02
Odor: Recognizable
Melting Point: 185-192°
Boiling Point: 130°/2mm
Beilstein Reference: 1459581
MDL No.: MFCD00007677
Formula:
C₆H₄Cl₂N₂O₂ Boiling Pt: 130 °C/2 mm Melting Pt: 185…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007677 CAS Number: 99-30-9 EINECS: 202-746-4 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...