(2-Pyridyl)acetic acid ethyl ester 98%

Supplier: Thermo Scientific Chemicals
Warning

B23988-06 B23988-14 B23988-22
CAAAB23988-06EA 108.7 CAD
CAAAB23988-06 CAAAB23988-14 CAAAB23988-22
(2-Pyridyl)acetic acid ethyl ester 98%
(2-Pyridyl)acetic acid ethyl ester

Purity: 98% CAS No.: 2739-98-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Flash Point: 110°(230°F) Refractive Index: 1.4970 Boiling Point: 69-70°/0.05mm Beilstein Reference: 123817 MDL No.: MFCD00006359

Formula: C₉H₁₁NO₂
Boiling Pt: 69…70 °C/0.05 mm
Melting Pt: 86…90 °C
Density: 1.084
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006359
CAS Number: 2739-98-2
EINECS: 220-365-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR