1-Bromo-4-butylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Bromophenyl)butane
B23979-14
B23979-22
CAAAB23979-14EA
120.4
CAD
CAAAB23979-14
CAAAB23979-22
CAAAB23979-36
1-Bromo-4-butylbenzene 98%
1-Bromo-4-butylbenzene
Purity: 98%
CAS No.: 41492-05-1
Molecular Formula: C10H13Br
Formula Weight: 213.12
Flash Point: 108°(226°F)
Refractive Index: 1.5300
Boiling Point: 242°
Beilstein Reference: 2080263
MDL No.: MFCD00040934
Formula:
C₁₀H₁₃Br Boiling Pt: 242 °C Melting Pt: –26 °C Density: 1.208 Flash Pt: 108 °C(226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040934 CAS Number: 41492-05-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...