Triacetoxymethylsilane 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Triacetoxy(methyl)silane, Methylsilanetriyl triacetate
B23974-09
B23974-18
B23974-30
CAAAB23974-09EA
64.15
CAD
CAAAB23974-09
CAAAB23974-18
CAAAB23974-30
Triacetoxymethylsilane 96%
Triacetoxymethylsilane
Purity: 96%
CAS No.: 4253-34-3
Molecular Formula: C7H12O6Si
Formula Weight: 220.25
Flash Point: 85°(185°F)
Melting Point: 40-45°
Boiling Point: 87-88°/3mm
Beilstein Reference: 1788668
UN No.: 1759
MDL No.: MFCD00008694
Formula:
C₇H₁₂O₆Si MW: 220.25 g/mol Boiling Pt: 87…88 °C/3 mm Melting Pt: 40…45 °C Density: 1.175 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008694 CAS Number: 4253-34-3 EINECS: 224-221-9 UN: 1759 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...