2-Hydroxy-1-naphthaldehyde 98%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Hydroxy-α-naphthaldehyde
B23951-14
B23951-22
CAAAB23951-14EA
57.51
CAD
CAAAB23951-14
CAAAB23951-22
2-Hydroxy-1-naphthaldehyde 98%, Technical Grade
2-Hydroxy-1-naphthaldehyde
Purity: 98%
CAS No.: 708-06-5
Molecular Formula: C11H8O2
Formula Weight: 172.18
Odor: Odorless
Melting Point: 77-80°
Boiling Point: 191-193°/27mm
UN No.: 3077
MDL No.: MFCD00004005
Formula:
C₁₁H₈O₂ MW: 172.18 g/mol Boiling Pt: 191…193 °C/27 mm Melting Pt: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004005 CAS Number: 708-06-5 EINECS: 211-902-0 UN: 3077 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...