1-Iodo-4-nitrobenzene 98+%

Supplier: Thermo Scientific Chemicals
Warning

B23950-06 B23950-14 B23950-22
CAAAB23950-06EA 73.94 CAD
CAAAB23950-06 CAAAB23950-14 CAAAB23950-22
1-Iodo-4-nitrobenzene 98+%
1-Iodo-4-nitrobenzene

Purity: 98+% CAS No.: 636-98-6 Molecular Formula: C6H4INO2 Formula Weight: 249.01 Melting Point: 172-177° Boiling Point: 288-290° MDL No.: MFCD00007299

Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 288…290 °C
Melting Pt: 172…177 °C
Density: 2.273 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007299
CAS Number: 636-98-6
EINECS: 211-272-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR