2,3-Dimethylphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23942-03
B23942-06
B23942-14
CAAAB23942-03EA
42.5
CAD
CAAAB23942-03
CAAAB23942-06
CAAAB23942-14
2,3-Dimethylphenylboronic acid 98%
2,3-Dimethylphenylboronic acid
Purity: 98%
CAS No.: 183158-34-1
Molecular Formula: C8H11BO2
Formula Weight: 149.99
Melting Point: ca 175°
Beilstein Reference: 8050763
Formula:
C₈H₁₁BO₂ Melting Pt: ∼ 175 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863524 CAS Number: 183158-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...