6-Methylquinaldine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23934-06
B23934-14
CAAAB23934-06EA
82.48
CAD
CAAAB23934-06
CAAAB23934-14
6-Methylquinaldine 98%
6-Methylquinaldine
Purity: 98%
CAS No.: 877-43-0
Molecular Formula: C11H11N
Formula Weight: 157.21
Melting Point: 56-58°
Boiling Point: 266-267°
MDL No.: MFCD00006762
Formula:
C₁₁H₁₁N Boiling Pt: 266…267 °C Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006762 CAS Number: 877-43-0 EINECS: 212-891-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...