3-Phenylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

B23930-06 B23930-14
CAAAB23930-06EA 93.69 CAD
CAAAB23930-06 CAAAB23930-14
3-Phenylpyridine 97%
3-Phenylpyridine

Purity: 97% CAS No.: 1008-88-4 Molecular Formula: C11H9N Formula Weight: 155.20 Flash Point: 110°(230°F) Refractive Index: 1.6160 Boiling Point: 272° Beilstein Reference: 110400 MDL No.: MFCD00006380

Formula: C₁₁H₉N
Boiling Pt: 272 °C
Melting Pt: 76…77 °C
Density: 1.085
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006380
CAS Number: 1008-88-4
EINECS: 213-762-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR