Methyl nicotinate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23908-30
B23908-22
B23908-36
CAAAAB23908-30EA
91.32
CAD
CAAAAB23908-30
CAAAB23908-22
CAAAB23908-36
Methyl nicotinate 99%
Methyl nicotinate
Purity: 99%
CAS No.: 93-60-7
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 95°(203°F)
Odor: Slightly sweetish
Boiling Point: 208-210°
Beilstein Reference: 113951
Merck Reference: 14,6100
MDL No.: MFCD00006388
Formula:
C₇H₇NO₂ Boiling Pt: 208…210 °C Melting Pt: 40…44 °C Density: 1.0048 g/cm³ (20 °C) Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006388 CAS Number: 93-60-7 EINECS: 202-261-8 Merck Index: 14,6100 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...