1,3-Diisopropylbenzene 96%
Supplier: Thermo Scientific Chemicals
Warning
|
B23904-18
B23904-30
CAAAB23904-18EA
59.25
CAD
CAAAB23904-18
CAAAB23904-30
1,3-Diisopropylbenzene 96%
1,3-Diisopropylbenzene
Purity: 96%
CAS No.: 99-62-7
Molecular Formula: C12H18
Formula Weight: 162.27
Flash Point: 76°(170°F)
Refractive Index: 1.4890
Melting Point: -63°
Boiling Point: 204°
Beilstein Reference: 1905828
MDL No.: MFCD00008889
Formula:
C₁₂H₁₈ MW: 162.27 g/mol Boiling Pt: 203 Melting Pt: -40 Density: 0.856 Flash Pt: 76 °C(170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008889 CAS Number: 99-62-7 EINECS: 202-773-1 UN: 3082 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...