3,5-Dibromophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

B23863-06 B23863-14
CAAAB23863-03EA 48.84 CAD
CAAAB23863-03 CAAAB23863-06 CAAAB23863-14
3,5-Dibromophenylboronic acid 97%
3,5-Dibromophenylboronic acid

Purity: 97% CAS No.: 117695-55-3 Molecular Formula: C6H5BBr2O2 Formula Weight: 279.73 Melting Point: 300° Beilstein Reference: 6645285

Formula: C₆H₅BBr₂O₂
MW: 279.72 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01075725
CAS Number: 117695-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR