1,3,5-Tribromobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23860-14
B23860-22
B23860-36
CAAAB23860-14EA
89.43
CAD
CAAAB23860-14
CAAAB23860-22
CAAAB23860-36
1,3,5-Tribromobenzene 98%
1,3,5-Tribromobenzene
Purity: 98%
CAS No.: 626-39-1
Molecular Formula: C6H3Br3
Formula Weight: 314.80
Melting Point: 121-124°
Boiling Point: 271°
MDL No.: MFCD00000080
Formula:
C₆H₃Br₃ Boiling Pt: 271 °C Melting Pt: 121…124 °C Density: 2.637 g/cm³ (22 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000080 CAS Number: 626-39-1 EINECS: 210-947-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...