1-Bromo-2-methylpropane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
B23854-0B
B23854-30
CAAAB23854-0BEA
270.18
CAD
CAAAB23854-0B
CAAAB23854-30
1-Bromo-2-methylpropane 98+%
1-Bromo-2-methylpropane
MDL: MFCD00000217
Beilstein: 1730915
Slightly soluble in water. Miscible with alcohol, ether
Purity: 98+%
CAS No.: 78-77-3
Molecular Formula: C4H9Br
Formula Weight: 137.02
Flash Point: 18°(64°F)
Refractive Index: 1.4350
Melting Point: -118°
Boiling Point: 90-92°
Solubility: Slightly soluble in water. Miscible with alcohol, ether
Beilstein Reference: 1730915
Merck Reference: 14,5135
UN No.: 2342
MDL No.: MFCD00000217
Formula:
C₄H₉Br Boiling Pt: 90…92 °C Melting Pt: –118 °C Density: 1.255 Flash Pt: 18 °C (64 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000217 CAS Number: 78-77-3 EINECS: 201-141-2 UN: 2342 ADR: 3,II Merck Index: 14,5135 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...