Furan-2-boronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23842-03
B23842-06
B23842-14
CAAAB23842-03EA
107.12
CAD
CAAAB23842-03
CAAAB23842-06
CAAAB23842-14
Furan-2-boronic acid 97%
Furan-2-boronic acid
Purity: 97%
CAS No.: 13331-23-2
Molecular Formula: C4H5BO3
Formula Weight: 111.89
Storage Temperature: -30°C to -10°C
Melting Point: ca 120° dec.
Beilstein Reference: 112370
MDL No.: MFCD00799544
Formula:
C₄H₅BO₃ Melting Pt: ca 120 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00799544 CAS Number: 13331-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...