1-Adamantanemethanol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol
B23831-06
B23831-14
CAAAB23831-03EA
42.72
CAD
CAAAB23831-03
CAAAB23831-06
CAAAB23831-14
1-Adamantanemethanol 98%
1-Adamantanemethanol
Purity: 98%
CAS No.: 770-71-8
Molecular Formula: C11H18O
Formula Weight: 166.26
Melting Point: 115-118°
Beilstein Reference: 1853339
Formula:
C₁₁H₁₈O Boiling Pt: 253 °C (1013 hPa) Melting Pt: 115…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00074751 CAS Number: 770-71-8 EINECS: 212-225-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...