2,4-Difluorophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23821-06
B23821-14
CAAAB23821-03EA
30.4
CAD
CAAAB23821-03
CAAAB23821-06
CAAAB23821-14
2,4-Difluorophenylboronic acid 97%
2,4-Difluorophenylboronic acid
Purity: 97%
CAS No.: 144025-03-6
Molecular Formula: C6H5BF2O2
Formula Weight: 157.91
Melting Point: 247-250°
Beilstein Reference: 8616257
Formula:
C₆H₅BF₂O₂ MW: 157.91 Melting Pt: 247…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318998 CAS Number: 144025-03-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...