3-Toluenethiol 97%

Supplier: Thermo Scientific Chemicals
Danger

B23818-06 B23818-14
CAAAB23818-06EA 88.32 CAD
CAAAB23818-06 CAAAB23818-14 CAAAB23818-22
3-Toluenethiol 97%
3-Toluenethiol

Purity: 97% CAS No.: 108-40-7 Molecular Formula: C7H8S Formula Weight: 124.21 Flash Point: 72°(161°F) Refractive Index: 1.5762 Melting Point: -20° Boiling Point: 196° Beilstein Reference: 1851959 Merck Reference: 14,9325 UN No.: 2810 MDL No.: MFCD00004843

Formula: C₇H₈S
Boiling Pt: 196 °C
Melting Pt: < –20 °C
Density: 1.044
Flash Pt: 72 °C(161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004843
CAS Number: 108-40-7
EINECS: 203-578-4
UN: 2810
ADR: 6.1,III
Merck Index: 14,9325

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR