2-Piperidinoethanol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(2-Hydroxyethyl)piperidine, 1-Piperidineethanol
B23816-22
B23816-36
CAAAB23816-22EA
63.36
CAD
CAAAB23816-22
CAAAB23816-36
2-Piperidinoethanol 99%
2-Piperidinoethanol
Purity: 99%
CAS No.: 3040-44-6
Molecular Formula: C7H15NO
Formula Weight: 129.20
Flash Point: 83°(181°F)
Refractive Index: 1.4804
Odor: Amine-like
pH: 12.4
Melting Point: 16°
Boiling Point: 199-202°
Beilstein Reference: 103390
UN No.: 2735
MDL No.: MFCD00006512
Formula:
C₇H₁₅NO MW: 129.2 g/mol Boiling Pt: 199…202 °C Melting Pt: 16 °C Density: 0.977 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006512 CAS Number: 3040-44-6 EINECS: 221-244-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...