Tribenzylamine 99+%

Supplier: Thermo Scientific Chemicals
Warning

B23797-14 B23797-22
CAAAB23797-14EA 39.34 CAD
CAAAB23797-14 CAAAB23797-22
Tribenzylamine 99+%
Tribenzylamine

Purity: 99+% CAS No.: 620-40-6 Molecular Formula: (C6H5CH2)3N Formula Weight: 287.41 Flash Point: 203°(397°F) Odor: Weak Boiling Point: 230°/13mm Beilstein Reference: 2214682 MDL No.: MFCD00004773

Formula: N(CH₂C₆H₅)₃
Boiling Pt: 230 °C/13 mm
Melting Pt: 91…94 °C
Density: 0.9912 g/cm³ (95 °C)
Flash Pt: 203 °C(397 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004773
CAS Number: 620-40-6
EINECS: 210-638-3
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c45b19a7c75302421b18150c43e96e03402f56d&vid=389de7f2863502b374023e1a04841d39e89717ec&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR