1,2,4-Tribromobenzene 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23778-06
B23778-14
CAAAB23778-06EA
67.94
CAD
CAAAB23778-06
CAAAB23778-14
1,2,4-Tribromobenzene 95%
Purity: 95%
CAS No.: 615-54-3
Molecular Formula: C6H3Br3
Formula Weight: 314.80
Flash Point:
Formula:
C₆H₃Br₃ Boiling Pt: 274…276 °C Melting Pt: 41…43 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000062 CAS Number: 615-54-3 EINECS: 210-433-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...