L(+)-Leucinol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
B23745-06
B23745-14
CAAAB23745-06EA
128.93
CAD
CAAAB23745-06
CAAAB23745-14
L(+)-Leucinol 97%
L(+)-Leucinol
Purity: 97%
CAS No.: 7533-40-6
Molecular Formula: C6H15NO
Formula Weight: 117.19
Flash Point: 90°(194°F)
Refractive Index: 1.4510
Boiling Point: 198-200°
Beilstein Reference: 1719240
Optical Rotation: +1.1° (neat)
MDL No.: MFCD00063676
Formula:
C₆H₁₅NO Boiling Pt: 198…200 °C Melting Pt: 66…70 °C Density: 0.917 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063676 CAS Number: 7533-40-6 EINECS: 231-400-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...