2,5-Dimethylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

B23740-03 B23740-06 B23740-14
CAAAB23740-03EA 43.61 CAD
CAAAB23740-03 CAAAB23740-06 CAAAB23740-14
2,5-Dimethylphenylboronic acid 98%
2,5-Dimethylphenylboronic acid

Purity: 98% CAS No.: 85199-06-0 Molecular Formula: C8H11BO2 Formula Weight: 149.99 Melting Point: ca 180° Beilstein Reference: 3241785

Formula: C₈H₁₁BO₂
Melting Pt: ∼ 180 °C
Storage Temperature: Ambient
MDL Number: MFCD01863525
CAS Number: 85199-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR