Trihexylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tri-n-hexylamine, Tri(hex-1-yl)amine

B23716-18 B23716-30
CAAAB23716-18EA 110.44 CAD
CAAAB23716-18 CAAAB23716-30
Trihexylamine 97%
Trihexylamine

Purity: 97% CAS No.: 102-86-3 Molecular Formula: [CH3(CH2)5]3N Formula Weight: 269.52 Flash Point: 144°(291°F) Refractive Index: 1.4415 Melting Point: -75° Boiling Point: 263-265° Beilstein Reference: 1755828 UN No.: 3082 MDL No.: MFCD00009523

Formula: C₁₈H₃₉N
Boiling Pt: 263…265 °C
Melting Pt: < –75 °C
Density: 0.798
Flash Pt: 144 °C(291 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009523
CAS Number: 102-86-3
EINECS: 203-062-9
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR