N,N'-Diphenyl-p-phenylenediamine 97%

Supplier: Thermo Scientific Chemicals
Warning

B23713-06 B23713-14 B23713-22
CAAAB23713-06EA 54.67 CAD
CAAAB23713-06 CAAAB23713-14 CAAAB23713-22
N,N'-Diphenyl-p-phenylenediamine 97%
N,N'-Diphenyl-p-phenylenediamine

Purity: 97% CAS No.: 74-31-7 Molecular Formula: C18H16N2 Formula Weight: 260.34 Melting Point: 146-153° Boiling Point: 220-225°/1mm Merck Reference: 14,3329 MDL No.: MFCD00003015

Formula: C₁₈H₁₆N₂
Boiling Pt: 220…225 °C/1 mm
Melting Pt: 146…153 °C
Density: 1.28
Storage Temperature: Ambient
MDL Number: MFCD00003015
CAS Number: 74-31-7
EINECS: 200-806-4
Merck Index: 14,3329

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR