2-Methyl-4-(trifluoromethoxy)aniline 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-5-(trifluoromethoxy)toluene, 4-Trifluoromethoxy-o-toluidine, 4-(Trifluoromethoxy)-2-toluidine

B23706-03 B23706-06
CAAAB23706-03EA 49.77 CAD
CAAAB23706-03 CAAAB23706-06
2-Methyl-4-(trifluoromethoxy)aniline 97+%
2-Methyl-4-(trifluoromethoxy)aniline

Purity: 97+% CAS No.: 86256-59-9 Molecular Formula: C8H8F3NO Formula Weight: 191.15 UN No.: 2810 MDL No.: MFCD01631541

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631541
CAS Number: 86256-59-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR