4-Chlorobenzhydrol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Chlorophenyl phenyl carbinol, 4-Chlorobenzhydryl alcohol, 4-Chloro-α-phenylbenzyl alcohol, p-Chlorobenzhydrol, 4-Chlorodiphenylmethanol

B23691-06 B23691-14 B23691-22
CAAAB23691-06EA 65.57 CAD
CAAAB23691-06 CAAAB23691-14 CAAAB23691-22
4-Chlorobenzhydrol 98%
4-Chlorobenzhydrol

Purity: 98% CAS No.: 119-56-2 Molecular Formula: C13H11ClO Formula Weight: 218.68 Melting Point: 58-62° Boiling Point: 150-151°/2.6mm MDL No.: MFCD00004491

Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
Boiling Pt: 150…151 °C/2.6 mm
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00004491
CAS Number: 119-56-2
EINECS: 204-333-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR