2-Phenylindole 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Phenyl-1H-indole

B23674-18 B23674-30
CAAAB23674-18EA 49.14 CAD
CAAAB23674-18 CAAAB23674-30
2-Phenylindole 95%
2-Phenylindole

Purity: 95% CAS No.: 948-65-2 Molecular Formula: C14H11N Formula Weight: 193.25 Melting Point: 189-190° Boiling Point: 250°/10mm MDL No.: MFCD00005608

Formula: C₁₄H₁₁N
MW: 193.25 g/mol
Boiling Pt: 250 °C/10 mm
Melting Pt: 189…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00005608
CAS Number: 948-65-2
EINECS: 213-436-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR