Butyl vinyl ether 98% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Butyl vinyl ether

Stabilization: 0.01% Stabilised with potassium hydroxide

B23673-AE B23673-AP
CAAAB23673-AEEA 54.19 CAD
CAAAB23673-AE CAAAB23673-AP
Butyl vinyl ether 98% stabilised
Butyl vinyl ether

Purity: 98% CAS No.: 111-34-2 Molecular Formula: C6H12O Formula Weight: 100.16 Storage Temperature: 2°C to 8°C Flash Point: -12°(10°F) Refractive Index: 1.4010 Melting Point: -113° Boiling Point: 92-94° Beilstein Reference: 1560217 UN No.: 3271 MDL No.: MFCD00009454

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 94
Melting Pt: -91.9
Density: 0.778
Flash Pt: –12 °C (10 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009454
CAS Number: 111-34-2
EINECS: 203-860-7
UN: 2352
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR