5-Bromo-2,2-difluoro-1,3-benzodioxole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-2,2-difluorobenzodioxole, 5-bromo-2,2-difluoro-2H-1,3-benzodioxole
B23661-14
B23661-06
CAAAAB23661-14EA
703.1
CAD
CAAAAB23661-14
CAAAB23661-03
CAAAB23661-06
5-Bromo-2,2-difluoro-1,3-benzodioxole 97%
5-Bromo-2,2-difluoro-1,3-benzodioxole
Purity: 97%
CAS No.: 33070-32-5
Molecular Formula: C7H3BrF2O2
Formula Weight: 237.00
Refractive Index: 1.4967
Boiling Point: 78-79°/20mm
MDL No.: MFCD00236212
Formula:
C₇H₃BrF₂O₂ MW: 237 g/mol Boiling Pt: 78…79 °C/20 mm Density: 1.740 Storage Temperature: Ambient |
MDL Number:
MFCD00236212 CAS Number: 33070-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...